6-Bromo-3-fluoro-2-methoxybenzeneboronic acid 98%

Supplier: Apollo Scientific

Synonyms: 2-Borono-3-bromo-6-fluoroanisole, (6-Bromo-3-fluoro-2-methoxyphenyl)boronic acid

PC7703-1G
APOSPC7703-1GEA 69.6 EUR
APOSPC7703-1G
6-Bromo-3-fluoro-2-methoxybenzeneboronic acid 98%
6-Bromo-3-fluoro-2-methoxybenzeneboronic acid
Formula: C₇H₇BBrFO₃
Storage Temperature: Ambient
MDL Number: MFCD09475867
CAS Number: 957035-08-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR