4-{4-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]piperazin-1-yl}benzonitrile 97%

Supplier: Apollo Scientific

PC8423-500MG
APOSPC8423-500MGEA 688 EUR
APOSPC8423-500MG
4-{4-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]piperazin-1-yl}benzonitrile 97%
4-{4-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]piperazin-1-yl}benzonitrile
Formula: C₁₇H₁₄ClF₃N₄
MW: 366.768 g/mol
Melting Pt: 144…146 °C
Storage Temperature: Ambient
MDL Number: MFCD00141702
CAS Number: 338979-20-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR