2-{2-[3-Fluoro-4-(trifluoromethyl)phenyl]-1,3-thiazol-4-yl}acetic acid

Supplier: Apollo Scientific

PC8591-1G PC8591-5G
APOSPC8591-1GEA 538 EUR
APOSPC8591-1G APOSPC8591-5G
2-{2-[3-Fluoro-4-(trifluoromethyl)phenyl]-1,3-thiazol-4-yl}acetic acid
2-{2-[3-Fluoro-4-(trifluoromethyl)phenyl]-1,3-thiazol-4-yl}acetic acid
Formula: C₁₂H₇F₄NO₂S
Melting Pt: 116…118 °C
Storage Temperature: Ambient
MDL Number: MFCD09027135
CAS Number: 937602-42-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR