Ethyl 2-(5-fluoropyridin-3-yl)acetate 95%

Supplier: Apollo Scientific

PC905564-1G PC905564-250MG
APOSPC905564-1GEA 806 EUR
APOSPC905564-1G APOSPC905564-250MG
Ethyl 2-(5-fluoropyridin-3-yl)acetate 95%
Ethyl-2-(5-fluoropyridin-3-yl)acetate
Formula: C₉H₁₀FNO₂
Storage Temperature: Ambient
MDL Number: MFCD16748979
CAS Number: 39891-14-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR