2,3-Difluoro-6-methylpyridine 98%

Supplier: Apollo Scientific

Synonyms: 2,3-Difluoro-6-picoline

PC906336-1G PC906336-5G
APOSPC906336-1GEA 101 EUR
APOSPC906336-1G APOSPC906336-5G
2,3-Difluoro-6-methylpyridine 98%
2,3-Difluoro-6-methylpyridine
Formula: C₆H₅F₂N
Storage Temperature: Ambient
MDL Number: MFCD16610828
CAS Number: 1227579-04-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR