Sodium hydroxide ≥98.0% (by acidimetry), pellets ACS, ISO for analysis

Supplier: PanReac AppliChem
Danger

Synonyms: Caustic soda

131687.0416 131687.1210 131687.1211 131687.1214
APPC131687.0416EA 584 EUR
APPC131687.0416 APPC131687.1210 APPC131687.1211 APPC131687.1214
Sodium hydroxide ≥98.0% (by acidimetry), pellets ACS, ISO for analysis
Sodium hydroxide

SOLUBILITY: water 1,090 g/l at 20 °C
PHYSICAL DESCRIPTION: White solid as lentils or flakes, frosted, caustic deliquescent, absorbing CO2 from the air

Formula: NaOH
Boiling Pt: 1390 °C
Melting Pt: 318 °C
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8,II

Order Now

Specification Test Results

Minimum Assay (Acidim.) 98.0%
Insoluble Matter in H₂O Max. 0.005%
Chloride (Cl) Max. 0.001%
Nitrogen Compounds (as N) Max. 0.0005%
Phosphate (PO₄) Max. 0.0005%
Sulphate (SO₄) Max. 0.002%
Silicate SiO₂ Max. 0.001%
Carbonate (as Na₂CO₃) Max. 1.0%
Heavy Metals (as Pb) Max. 0.0005%
Al Max. 0.0005%
As Max. 0.00004%
Ca Max. 0.0005%
Cd Max. 0.0005%
Co Max. 0.0005%
Cu Max. 0.0005%
Fe Max. 0.0005%
Hg Max. 0.00001%
K Max. 0.02%
MG Max. 0.0005%
Mn Max. 0.0005%
Ni Max. 0.0005%
PB Max. 0.0005%
Zn Max. 0.001%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR