Sulphuric acid 98% 98 determination_nitrogen

Supplier: PanReac AppliChem

173163.1611 173163.1612
APPC173163.1611EA 45.8 EUR
APPC173163.1611 APPC173163.1612
Sulphuric acid 98% 98 determination_nitrogen
Sulphuric acid
Formula: H₂SO₄
MW: 98.08 g/mol
Boiling Pt: 330 °C
Melting Pt: –15 °C
Density: 1.84
MDL Number: MFCD00064589
CAS Number: 7664-93-9

Order Now

Specification Test Results

Assay 98%
Appearence Clear colorless liquid
Density 20/4 1,84
Residue on ignition 0,0005%
Nitrogen compounds (as N) 0,0005%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR