Sodium hydroxide 10 mol/l (10 N) volumetric solution
Supplier: PanReac AppliChem
Danger
Synonyms: Caustic soda
183508.1211
APPC183508.1211EA 48 EUR
APPC183508.1211
Sodium hydroxide 10 mol/l (10 N) volumetric solution
Sodium hydroxide
Certifications: Traceable to NIST.
Formula: NaOH
Density: 1.331 kg/l
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II
Order Now
Specification Test Results
Titer at 20 °C 0.99 - 1.01
Uncertainty See certificate
Traceability NIST