Sodium hypochlorite solution 5% (w/w), Technical Grade

Supplier: PanReac AppliChem
Danger

212297.1214 212297.1211
APPC212297.1214EA 45.4 EUR
APPC212297.1214 APPC212297.1211
Sodium hypochlorite solution 5% (w/w), Technical Grade
Sodium hypochlorite
Formula: ClNaO
MW: 74.44 g/mol
Melting Pt: -6
Storage Temperature: Ambient
MDL Number: MFCD00011120
CAS Number: 7681-52-9
UN: 1791
ADR: 8 III
Merck Index: 13,08702

Order Now

Specification Test Results

Assay (iodometr.) 5% ± 0.5% (At the moment of the batch analysis)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR