Sodium hypochlorite solution 13% (w/v), Technical Grade

Supplier: PanReac AppliChem

213322.1214 213322.0715
APPC213322.1214EA 33 EUR
APPC213322.1214 APPC213322.0715
Sodium hypochlorite solution 13% (w/v), Technical Grade
Sodium hypochlorite
Formula: ClNaO
MW: 74.44 g/mol
Melting Pt: -6
Storage Temperature: Ambient
MDL Number: MFCD00011120
CAS Number: 7681-52-9
UN: 1791
ADR: 8 III
Merck Index: 13,08702

Order Now

Specification Test Results

Assay (Active Chlorine, Iodometry) 10.0 - 14.0%
NaCl Max. 11.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR