Davidson solution

Supplier: PanReac AppliChem

A3200.1000
APPCA3200.1000EA 197 EUR
APPCA3200.1000
Davidson solution
Stains Microscopy and staining reagents

Davidson solution is a rapid fixative.


Composition:
Deionised water: 347 ml/l
Acetic acid 100 %: 111 ml/l
Ethanol 99 %: 320 ml/l
Formaldehyde - Solution 10 % phosphate buffered: 222 ml/l

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR