2-Iodobutane ≥99% stabilised

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 2-Butyl iodide, sec-Butyl iodide

Stabilization: Stabilised with copper

B20619.18 B20619.30 B20619.36
B20619.18EA 40.1 EUR
B20619.18 B20619.30 B20619.36
2-Iodobutane ≥99% stabilised
2-Iodobutane

Stabilized with copper


Purity: 99% CAS No.: 513-48-4 Molecular Formula: C4H9I Formula Weight: 184.02 Flash Point: 21°(70°F) Refractive Index: 1.4991 Odour: Ether-like Melting Point: -104° Boiling Point: 119-120° Beilstein Reference: 1718777 Merck Reference: 14,1574 UN No.: 2390 MDL No.: MFCD00001072

Formula: C₄H₉I
Boiling Pt: 119…120 °C
Melting Pt: –104 °C
Density: 1.597
Flash Pt: 21 °C (70 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001072
CAS Number: 513-48-4
EINECS: 208-163-1
UN: 2390
ADR: 3,II
Merck Index: 14,1574

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR