5-Iodoindole ≥98%

Supplier: Thermo Fisher Scientific
Warning

B20638.06 B20638.14
B20638.06EA 96.4 EUR
B20638.06 B20638.14
5-Iodoindole ≥98%
5-Iodoindole

Purity: 98% CAS No.: 16066-91-4 Molecular Formula: C8H6IN Formula Weight: 243.05 Melting Point: 97-100° MDL No.: MFCD00220065

Formula: C₈H₆IN
Melting Pt: 97…100 °C
Storage Temperature: Ambient
MDL Number: MFCD00220065
CAS Number: 16066-91-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR