2-(3-Phenylpropyl)pyridine ≥99%

Supplier: Thermo Fisher Scientific
Warning

B22495.03 B22495.06 B22495.14
B22495.03EA 30.4 EUR
B22495.03 B22495.06 B22495.14
2-(3-Phenylpropyl)pyridine ≥99%
2-(3-Phenylpropyl)pyridine

Purity: 99% CAS No.: 2110-18-1 Molecular Formula: C14H15N Formula Weight: 197.28 Flash Point: 100°(212°F) Boiling Point: 142-143°/1mm MDL No.: MFCD00191237

Formula: C₁₄H₁₅N
Boiling Pt: 142…143 °C (1 mmHg)
Density: 1.017
Flash Pt: 100 °C (212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00191237
CAS Number: 2110-18-1
EINECS: 218-300-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR