Ammonium 1-pyrrolidinecarbodithioate ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Pyrrolidinedithiocarbamate ammonium salt, 1-Pyrrolidinecarbodithioic acid, ammonium salt, Ammonium pyrrolidine-1-carbodithioate, Pyrrolidine-1-Dithiocarboxylic Acid Ammonium Salt, APCD

B23731.14 B23731.22
B23731.14EA 48.3 EUR
B23731.14 B23731.22
Ammonium 1-pyrrolidinecarbodithioate ≥98%
Ammonium 1-pyrrolidinecarbodithioate

Purity: 98% CAS No.: 5108-96-3 Molecular Formula: C5H12N2S2 Formula Weight: 164.29 Melting Point: 150-155° Beilstein Reference: 3730472

Formula: C₅H₁₂N₂S₂
Melting Pt: 150…155 °C
Storage Temperature: Ambient
MDL Number: MFCD00012720
CAS Number: 5108-96-3
EINECS: 225-834-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR