Ethyl cyclopropanecarboxylate ≥98%

Supplier: Thermo Fisher Scientific
Danger

B24870.14 B24870.22
B24870.14EA 49.5 EUR
B24870.14 B24870.22
Ethyl cyclopropanecarboxylate ≥98%
Ethyl cyclopropanecarboxylate

Purity: 98% CAS No.: 4606-07-9 Molecular Formula: C6H10O2 Formula Weight: 114.15 Flash Point: 18°(64°F) Refractive Index: 1.4200 Boiling Point: 133-134° Beilstein Reference: 2039752 UN No.: 3272 MDL No.: MFCD00001282

Formula: C₆H₁₀O₂
Boiling Pt: 133…134 °C
Density: 0.960
Flash Pt: 18 °C (64 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001282
CAS Number: 4606-07-9
EINECS: 225-010-4
UN: 3272
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR