N-Benzylidenemethylamine ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: N-Benzylidenemethanamine

B25161.06 B25161.14 B25161.22
B25161.06EA 52.8 EUR
B25161.06 B25161.14 B25161.22
N-Benzylidenemethylamine ≥98%
N-Benzylidenemethylamine

Purity: 98+% CAS No.: 622-29-7 Molecular Formula: (C6H5)CH=NCH3 Formula Weight: 119.17 Storage Temperature: 2°C to 8°C Flash Point: 77°(170°F) Refractive Index: 1.5520 Boiling Point: 66-69°/10mm Beilstein Reference: 878278 MDL No.: MFCD00008294

Formula: C₈H₉N
Boiling Pt: 66…69 °C (10 mmHg)
Density: 0.967
Flash Pt: 77 °C (170 °F)
MDL Number: MFCD00008294
CAS Number: 622-29-7
EINECS: 210-728-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR