SONOREX DIGITEC DT 514, ultrasonic bath, 230 V( 10%) 50/60 Hz, ID 325x300x150 mm 13,5 l, s/s digital display timer 1, 2, 3, 4, 5, 10, 15,30 min and continuous, ultrasonic peak output 860 W, HF output 215 Weff., SweepTec and DEGAS, drain: ball va 1 * 1 items
Supplier: BANDELIN-ELECTRONIC
SONOREX DIGITEC DT 514, ultrasonic bath, 230 V( 10%) 50/60 Hz, ID 325x300x150 mm 13,5 l, s/s digital display timer 1, 2, 3, 4, 5, 10, 15,30 min and continuous, ultrasonic peak output 860 W, HF output 215 Weff., SweepTec and DEGAS, drain: ball va 1 * 1 items