Digital heating shaking block heater

Supplier: Thermo Fisher Scientific
88880028
BARN88880028EA 1580 EUR
BARN88880028
Digital heating shaking block heater
Block Heaters Shaking Block Heaters
This can be used as a drybath, shaker or both. The wide temperature range covers a broad range of applications including thawing, incubation, enzyme reactions, blood urea nitrogen determinations, and immunoassays.

  • PID control ensures consistent shaking results and precise temperature control
  • Stores ten programs and up to ten steps per program
  • Interval shaking function allows shaking and nonshaking in same program
  • Cord set with various plug types
  • Low-profile design
  • Easily changed blocks in multiple sizes for maximum flexibility

Ordering information: More Product Information
Order Now

SPECIFICATIONS

Model Digital shaking block heater
Environmentally Preferable
Orbit 2 mm
Temperature control range Ambient +5...100 °C
Temperature accuracy ±0,5 °C @ 80 °C
No. of blocks 1
W×D×H 76×334×170 mm
Weight 8,3 kg
Speed range 150 - 1500 min⁻¹ (96-well block);
150 - 1200 min⁻¹ (0,5, 1,5 and 50 ml tube blocks)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR