Aerosol evacuation system for CytoFLEX SRT
Supplier: Beckman Coulter
C63648
BECMC63648EA 13560 EUR
BECMC63648
Aerosol evacuation system for CytoFLEX SRT
Cell Counters and Flow Cytometers Flow Cytometers
The Aerosol Evacuation System removes aerosols and micro droplets, which may be generated during the course of normal operation or sort failure conditions, from the sort chamber without disturbing sorting.
  • Ultra-quiet motor (40 dB Min and 69,5 dB Max) and a high-suction, high flow rate centrifugal action pump
  • Micro droplets and particulates greater than 0,12 μm are removed under vacuum and trapped in an Ultra-Low Penetration Air (ULPA) filter
  • Air flow rate is fully user adjustable, providing clearance of the sort chamber at rates of 10 to 40 complete air