Single-channel electronic timer with triple alarms and certificate of calibration, SP Bel-Art H-B® DURAC®
Supplier: Bel-Art Products, a Part of SP
B617003600
BELAB617003600EA 143 EUR
BELAB617003600
Single-channel electronic timer with triple alarms and certificate of calibration, SP Bel-Art H-B® DURAC®
Timers Digital Timers
Choice of vibration, sound, and light alarms makes this timer ideal for use in high noise locations.
  • Timing capacity: 99 minutes, 59 seconds
  • Count direction: up/down
  • Vibration, light, and loud buzzer alarms can function at one time, or be independently shut off
  • Clock function
  • Timer features a certificate of calibration and individual serial number
Lay flat on a bench, use the flip-open stand for a vertical view, or carry on a lab coat with the pocket clip.
Display Dimensions: 35×20 mm.
Certifications: CE marked. RoHS compliant. These electronicMore Product Information