Gas dispersion tube, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
F136910000
BELAF136910000EA 142 EUR
BELAF136910000
Gas dispersion tube, SP Bel-Art
Gas Tubes
HDPE gas dispersion tube. Bell-mouthed fitting with a sealed-in, coarse porosity Fritware® PE disc at the large end.
  • Smaller bubbles
  • Tube internal diameter: 6,35 mm
Suitable for insertion into small openings.