Scrubbing sponge for glassware, Cleanware™, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
F170780001
BELAF170780001EA 48.8 EUR
BELAF170780001
Scrubbing sponge for glassware, Cleanware™, SP Bel-Art
Wipes Sponges
This odor-resistant, highly absorbent sponge is designed to remove stubborn residue on glassware.
  • Wipe dry performance equal to cellulose; maintains the desirable physical properties of polyurethane
  • Foam contains anti-microbial technology to inhibit odor causing bacteria within the sponge itself
  • Stays soft and flexible, wet or dry; will not crack or shrink, or become brittle like cellulose
  • Scouring side resists snagging; does not leave fibers behind and retains aggressive scouring ability even with frequent use