Desiccant replacement beads, Secador®, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
F420740020
BELAF420740020EA 53.8 EUR
BELAF420740020
Desiccant replacement beads, Secador®, SP Bel-Art
Desiccators Desiccants
These colour-indicating silica gel beads are for use in refilling the Secador® auto-desiccator module which automatically regenerates them for thousands of cycles.
  • Silica gel absorbs moisture from the air and is inert, non toxic and non corrosive
  • Beads turn from blue to light pink indicating that the silica gel is saturated and cannot absorb additional moisture
  • Beads turn pink/dark brown and fail to regain their blue colour when it is time to replace the beads with a fresh supply