Fume hood, benchtop, small, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
H500000002
BELAH500000002EA 1170 EUR
BELAH500000002
Fume hood, benchtop, small, SP Bel-Art
Fume Hoods and Enclosures Pharmacy Fume Hoods
Benchtop fume hood designed for small facilities or installations in which a number of fume hoods are needed, this unit is mainly used in schools, junior colleges and universities.
  • Easy to clean
  • Bottom gap allows for continuous air flow
  • One-piece shell moulded of chemical resistant polyethylene with rounded inside corners
  • Clear acrylic sash slides on a track featuring stop pins for open, closed or intermediate positions