Bottle, capacity 2l, body diameter 136mm, body height 260mm, with red cap/pouring ring, thread size 45mm, printed with retrace code: downloadable batch certificate, withstands hot air sterilisation up to 180°C, PTFE sealing disc resists chemical att 1 * 10 items
Supplier: SciLabware