Ductless fume hoods, Chemcap™ Clearview™
Supplier: BIGNEAT
BC0603-4 BC1206-4 BC1506-4
BIGNBC0603-4EA 7950 EUR
BIGNBC0603-4 BIGNBC1206-4 BIGNBC1506-4
Ductless fume hoods, Chemcap™ Clearview™
Fume Hoods and Enclosures Ductless Fume Hoods
Chemcap™ Clearview™ ductless fume hoods include Chemcap™ carbon filtration. A range of six standard cabinet sizes with keypad control panel, touch button controls and service due indicator. Includes 18 W fluorescent lamp and PVC base tray which offers excellent chemical resistance and contains spillages.
Cabinets are manufactured from polyester coated steel; transparent panels are acrylic and resistant to corrosion.
  • Fully portable
  • Clearview all-round vision
  • BC 4002 has a full width envelope access
  • BC 6003 to BC 1204 have teardrop front working access
  • BC 1206