Skeleton model, with hanging stand
Supplier: 3B Scientific
1020172
BINH1020172EA 1060 EUR
BINH1020172
Skeleton model, with hanging stand
Models Human Biology Models
Stan, the standard model of a human skeleton has a very high quality and robust construction along with a stable metal hanging stand.
  • Made from a durable, unbreakable synthetic material
  • Hanging stand with five casters (painted white)
  • 3 part assembled skull and individually inserted teeth
  • Limbs can be removed quickly and easily
Weight: 8,77 kg