Mouse Lipocalin-2 ELISA kit
Supplier: Biorbyt
ORB54813-96
BIRBORB54813-96EA 0 EUR
BIRBORB54813-96
Mouse Lipocalin-2 ELISA kit
Assays ELISAs
Mouse Lipocalin-2 ELISA kit for the quantification of mouse lipocalin-2 in serum, plasma, urine and cell culture supernate samples.
Lipocalin-2 (LCN2), also known as neutrophil gelatinase-associated lipocalin (NGAL), 24p3, or neutrophil lipocalin (NL), is a 25-kDa secretory glycoprotein. LCN2 has been implicated in a variety of cellular processes including the innate immune response, differentiation, tumorigenesis, and cell survival.