L(+)-Valine

Supplier: BIOMOL RESEARCH LABORATORIES

Synonyms: H-Val-OH, Valine, (S)-(+)-Valine

23770-1 23770-2
BMOL23770-1EA 0 EUR
BMOL23770-1 BMOL23770-2
L(+)-Valine
L(+)-Valine
Formula: C₅H₁₁NO₂
Boiling Pt: 217 °C (1013 hPa)
Melting Pt: 86 °C
Density: 1.284 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064220
CAS Number: 72-18-4
EINECS: 200-773-6
Merck Index: 13,09458

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR