N,N,N',N'-Tetramethylethylenediamine (TEMED)

Supplier: BIOMOL RESEARCH LABORATORIES
Danger

Synonyms: 1,2-Bis(dimethylamino)ethane, TMEDA, N,N,N',N'-Tetramethylethylenediamine

50406-1 50406-2
BMOL50406-1EA 0 EUR
BMOL50406-1 BMOL50406-2
N,N,N',N'-Tetramethylethylenediamine (TEMED)
N,N,N',N'-Tetramethylethylenediamine (TEMED)
Formula: C₆H₁₆N₂
Boiling Pt: 120 °C (1013 hPa)
Melting Pt: –64 °C
Density: 0.7765 g/cm³ (20 °C)
Flash Pt: 17 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008335
CAS Number: 110-18-9
EINECS: 203-744-6
UN: 2372
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6838f1f5b7d0f69dcc81a8ed247f6c178dad231c&vid=ea361367e9944e4101e38622fdd18b07756eb676&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR