Luminol (3-Aminophthalhydrazide)

Supplier: BIOMOL RESEARCH LABORATORIES

Synonyms: NSC 5064, Luminol, 5-Amino-2,3-Dihydro-1,4-Phthalazinedione, 5-Amino-2,3-dihydrophthalazine-1,4-dione

54570-2 54570-3
BMOL54570-2EA 0 EUR
BMOL54570-2 BMOL54570-3
Luminol (3-Aminophthalhydrazide)
Luminol (3-Aminophthalhydrazide)
Formula: C₈H₇N₃O₂
MDL Number: MFCD00006890
CAS Number: 521-31-3
EINECS: 208-309-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR