Anti-SPN Mouse Monoclonal Antibody (CF568) [clone: SPN/839]
Supplier: Biotium
BNC680839-500 BNC680839-100
BNC680839-500EA 560 EUR
BNC680839-500 BNC680839-100
Anti-SPN Mouse Monoclonal Antibody (CF568) [clone: SPN/839]
Antibodies
This antibody recognizes a cell surface glycoprotein of 95/115/135 kDa (depending upon the extent of glycosylation), identified as CD43. 70-90% of T-cell lymphomas and from 22-37% of B-cell lymphomas express CD43. No reactivity has been observed with reactive B-cells. So a B-lineage population that co-expresses CD43 is highly likely to be a malignant lymphoma, especially a low-grade lymphoma, rather than a reactive B-cell population. When CD43 antibody is used in combination with anti-CD20, effective immunophenotyping of the lymphomas in formalin-fixed tissues can be obtained. Co-staining ofMore Product Information