Anti-LHB Mouse Monoclonal Antibody (Purified) [clone: LHb/1214]
Supplier: Biotium
BNUB1214-500 BNUB1214-100
BNUB1214-500EA 511 EUR
BNUB1214-500 BNUB1214-100
Anti-LHB Mouse Monoclonal Antibody (Purified) [clone: LHb/1214]
Antibodies
Luteinizing hormone (LH) is a glycoprotein. Each monomeric unit is a sugar-like protein molecule; two of these make the full, functional protein. Its structure is similar to the other glycoproteins, follicle-stimulating hormone (FSH), thyroid-stimulating hormone (TSH), and human chorionic gonadotropin (hCG). The protein dimer contains 2 polypeptide units, labeled alpha and beta subunits that are connected by two bridges. The alpha subunits of LH, FSH, TSH, and hCG are identical, and contain 92 amino acids. The beta subunits vary. LH has a beta subunit of 121 amino acids (LHB) that confersMore Product Information