Drums
Supplier: BOCHEM
8301 8304 8305 8341 8342 8346 8351 8352 8356 8307 8348 8358 8300 8302 8347 8340 8350 8354 8357 8344
BOCH8301EA 320 EUR
BOCH8301 BOCH8304 BOCH8305 BOCH8341 216-0234 BOCH8346 BOCH8351 BOCH8352 BOCH8356 BOCH8307 BOCH8348 BOCH8358 BOCH8300 BOCH8302 BOCH8347 BOCH8340 BOCH8350 BOCH8354 BOCH8357 BOCH8344
Drums
Sample Containers General Purpose Containers
Made of 18/10 stainless steel.
  • With or without lid available
  • With handle
  • Very good temperature resistance