Baktolan® Lotion

Supplier: Bode
980171
BODL980171EA 144 EUR
BODL980171
Baktolan® Lotion
Skincare Skin Creams Hand and Body Lotions
Moisturising, fast-absorbing fragrance-free oil-in-water (o/w) lotion for normal skin.

  • Fragrance-free
  • Nurturing formula with plant-based oils and vitamin E
  • Increases significantly the skin's moisture content is quickly absorbed leaving no residues
  • Provides a smooth and soft skin feeling
  • Free of parabens
  • Ideal for the quick skin care in between
  • Appearance: White to slightly white emulsion
  • Density: 0,990 g/cm³
  • pH value: 5,5 to 6,5

Baktolan lotion pure is a fragrance-free light oil-in-water lotion matched to the skin's pH-value. Particular value wasMore Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR