Tissue float baths, deluxe
Supplier: BOEKEL SCIENTIFIC
145951-2
BOEI145951-2EA 2840 EUR
BOEI145951-2
Tissue float baths, deluxe
Baths Tissue Baths
Precision temperature controlled water bath that eliminates wrinkles and distortion during the preparation of paraffin embedded tissue sections.
  • Digital microprocessor control
  • LED with illuminated black background, transparent PYREX® dish allows specimen to be easily seen
  • Slide dryer block to assist in drying slides
  • Tissue orienter block to smooth wrinkled sections