Ground joint distributors, BOLA
Supplier: Bohlender
B302-02
BOHLB302-02EA 423 EUR
BOHLB302-02
Ground joint distributors, BOLA
Adapters Adapters, Conical Ground Joints
PTFE, white.
  • Temperature resistance: −200 to +250 °C
  • Universal chemical resistance
  • Autoclavable
With ground joint cone and socket size 29. All ground joints are connected with a bore dia. of 10 mm. The bore dia. of the cone is 16 mm. Integrated special nut (Safe-Lab) for easy locking and unlocking of the ground joint.