Multiple distributors for bottles, BOLA
Supplier: Bohlender
D651-04
BOHLD651-04EA 135 EUR
BOHLD651-04
Multiple distributors for bottles, BOLA
Closures Bottle Closures
Blue screw cap made of PPS. Body made of PTFE.
  • Temperature resistant from −20 to +200 °C
  • Chemical resistant
  • Autoclavable at 121 °C
FDA conform.