Anti-MUC16 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss
BS-0091R-HRP
BOSSBS-0091R-HRPEA 909 EUR
BOSSBS-0091R-HRP
Anti-MUC16 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Thought to provide a protective, lubricating barrier against particles and infectious agents at mucosal surfaces.

Recommended Dilutions: IHC-P: 1:100-500

Type: Primary
Antigen: MUC16
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR