Anti-TRKB Rabbit Polyclonal Antibody (Cy5®)
Supplier: Bioss
BS-0175R-CY5
BOSSBS-0175R-CY5EA 909 EUR
BOSSBS-0175R-CY5
Anti-TRKB Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Receptor tyrosine kinase involved in the development and the maturation of the central and the peripheral nervous systems through regulation of neuron survival, proliferation, migration, differentiation, and synapse formation and plasticity. Receptor for BDNF/brain-derived neurotrophic factor and NTF4/neurotrophin-4. Alternatively can also bind NTF3/neurotrophin-3 which is less efficient in activating the receptor but regulates neuron survival through NTRK2. Upon ligand-binding, undergoes homodimerization, autophosphorylation and activation. Recruits, phosphorylates and/or activates severalMore Product Information