Anti-ILK Rabbit Polyclonal Antibody (Cy7®)
Supplier: Bioss
BS-0317R-CY7
BOSSBS-0317R-CY7EA 900 EUR
BOSSBS-0317R-CY7
Anti-ILK Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Receptor-proximal protein kinase regulating integrin-mediated signal transduction. May act as a mediator of inside-out integrin signaling. Focal adhesion protein part of the complex ILK-PINCH. This complex is considered to be one of the convergence points of integrin- and growth factor-signaling pathway. Could be implicated in mediating cell architecture, adhesion to integrin substrates and anchorage-dependent growth in epithelial cells. Phosphorylates beta-1 and beta-3 integrin subunit on serine and threonine residues, but also AKT1 and GSK3B.
Recommended Dilutions: IF(IHC-P): 1:50-200;More Product Information