Anti-NIS Rabbit Polyclonal Antibody

Supplier: Bioss
BS-0448R
BOSSBS-0448REA 752 EUR
BOSSBS-0448R
Anti-NIS Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: NIS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR