Anti-BrdU Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss
BS-0489R-CY5.5
BOSSBS-0489R-CY5.5EA 883 EUR
BOSSBS-0489R-CY5.5
Anti-BrdU Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
The immunocytochemical detection of bromodeoxyuridine (BrdU) incorporated into DNA is a powerful tool to study the cytokinetics of normal and neoplastic cells. In vitro or in vivo labeling of tumor cells with the thymidine analogue BrdU and the subsequent detection of incorporated BrdU with specific anti-BrdU monoclonal is an accurate and comprehensive method to quantitate the degree of DNA-synthesis.BrdU is incorporated into the newly synthezised DNA of S-phase cells may provide an estimate for the fraction of cells in S-phase. Also dynamic proliferative information such as the S-phaseMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol BrdU
Antigen name Bromodeoxyuridine
Environmentally Preferable
Antibody type Primary
Clonality Polyclonal
Conjugation Cy5.5®
Reactivity Multiple
Host Rabbit
Isotype IgG
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR