Anti-PRL Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss
BS-0508R-A488
BOSSBS-0508R-A488EA 901 EUR
BOSSBS-0508R-A488
Anti-PRL Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
This secreted hormone is a growth regulator for many tissues, including cells of the immune system. It may also play a role in cell survival by suppressing apoptosis, and it is essential for lactation. Prolactin acts primarily on the mammary gland by promoting lactation.

Recommended Dilutions: IF(IHC-P): 1:50-200; IF(ICC)(1:50-200

Type: Primary
Antigen: PRL
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR