Anti-CDK5 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Supplier: Bioss
BS-0559R-A350
BOSSBS-0559R-A350EA 901 EUR
BOSSBS-0559R-A350
Anti-CDK5 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
CDK5 is serine/threonine kinase involved in synaptic regulation and neuronal development; phosphorylates synaptic protein Pctaire1; regulates acetylcholine receptor expression.CDK5 is a member of the cyclindependent kinase family of serine/threonine kinases. It is present in numerous mammalian tissues including kidney, testes, and ovary. Its activity is detected almost exclusively in brain extracts. Neuronal and muscle cells contain the highest amount of this protein. Similar to other Cdks, monomeric Cdk5 displays no enzymatic activity, but Cdk5 is not activated by cyclins. Instead, the p35More Product Information