Anti-BIRC7 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss
BS-0738R-CY5
BOSSBS-0738R-CY5EA 909 EUR
BOSSBS-0738R-CY5
Anti-BIRC7 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Apoptotic regulator capable of exerting proapoptotic and anti-apoptotic activities and plays crucial roles in apoptosis, cell proliferation, and cell cycle control. Its anti-apoptotic activity is mediated through the inhibition of CASP3, CASP7 and CASP9, as well as by its E3 ubiquitin-protein ligase activity. As it is a weak caspase inhibitor, its anti-apoptotic activity is thought to be due to its ability to ubiquitinate DIABLO/SMAC targeting it for degradation thereby promoting cell survival. May contribute to caspase inhibition, by blocking the ability of DIABLO/SMAC to disrupt XIAP/BIRC4-More Product Information
Order Now

SPECIFICATIONS

Antigen symbol BIRC7
Antigen name Baculoviral IAP Repeat Containing 7
Environmentally Preferable
Antigen synonyms MLIAP|Baculoviral IAP repeat-containing protein 7|Kidney inhibitor of apoptosis protein|Melanoma inhibitor of apoptosis protein|LIVIN|RING finger protein 50|RNF50|BIRC7|ML-IAP|KIAP
Antibody type Primary
Clonality Polyclonal
Conjugation Cy5®
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 79444
Isotype IgG
ImmunoChemistry Yes
Immunogen 100-150/298
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR