Anti-COL3A1 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss
BS-0948R-CY5.5
BOSSBS-0948R-CY5.5EA 901 EUR
BOSSBS-0948R-CY5.5
Anti-COL3A1 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
Collagen type III occurs in most soft connective tissues along with type I collagen. Involved in regulation of cortical development. Is the major ligand of GPR56 in the developing brain and binding to GPR56 inhibits neuronal migration and activates the RhoA pathway by coupling GPR56 to GNA13 and possibly GNA12.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: COL3A1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR