Anti-BPIFB1 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss
BS-0976R-CY3
BOSSBS-0976R-CY3EA 900 EUR
BOSSBS-0976R-CY3
Anti-BPIFB1 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
May play a role in innate immunity in mouth, nose and lungs. Binds bacterial lipopolysaccharide (LPS) and modulates the cellular responses to LPS.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BPIFB1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR