Anti-SPC Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Supplier: Bioss
BS-10067R-PE
BOSSBS-10067R-PEEA 944 EUR
BOSSBS-10067R-PE
Anti-SPC Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antibodies
Pulmonary surfactant associated proteins promote alveolar stability by lowering the surface tension at the air-liquid interface in the peripheral air spaces.

Recommended Dilutions: Flow Cytometry: 1:20-100

Type: Primary
Antigen: SPC
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR